AS601245 sum of isomers ≥95% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: (Z)-2-(benzo[d]thiazol-2-yl)-2-(2-((2-(pyridin-3-yl)ethyl)imino)-1,2-dihydropyrimidin-4-yl)acetonitrile, (2Z)-2-(3H-1,3-Benzothiazol-2-ylidene)-2-[2-(2-pyridin-3-ylethylamino)pyrimidin-4-yl]acetonitrile, SAPK Inhibitor V, 2-(Benzo[d]thiazol-2-yl)-2-(2-((2-(pyridin-3-yl)ethyl)amino)pyrimidin-4-yl)acetonitrile, 1,3-Benzothiazol-2-yl-(2-((2-(3-pyridinyl)ethyl)amino)-4-pyrimidinyl)acetonitrile

270-443-M001 270-443-M005 270-443-M025
89147-914EA 113.76 CAD
89147-914 89147-916 89147-918
AS601245 sum of isomers ≥95% (by HPLC)
AS601245

Jnk inhibitor


Potent and cell permeable ATP competitive JNK inhibitor (hJNK1: IC50=150nM, hJNK2: IC50=220nM and hJNK3: IC50=70 nM). Displays anti-inflammatory properties and has been shown to reduce TNF-α plasma levels induced by LPS in mice.

Formula: C₂₀H₁₆N₆S
MW: 372.45 g/mol
MDL Number: MFCD07772196
CAS Number: 345987-15-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2843e7bf1e1d4535bd2dbd47abfbf034a1148496&vid=01307e2cf6b828a9843af1a738278d997fd1568f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR