1-Azakenpaullone ≥95% (by NMR)

Supplier: Enzo Life Sciences

Synonyms: 9-Bromo-7,12-dihydropyrido[3',2':2,3]azepino[4,5-b]indol-6(5H)-one

270-430-M001 270-430-M005
89147-882EA 218.04 CAD
89147-882 89147-884
1-Azakenpaullone ≥95% (by NMR)
1-Azakenpaullone

GSK-3beta inhibitor


Analog of kenpaullone (Prod. No. BML-EI310) acting as a potent and ATP-competitive inhibitor of GSK-3beta (IC50=18nM). Displays ~100-200 fold greater selectivity on GSK-3beta than over CDK1/cyclin B and CDK5/p25 (IC50=2.0µM and 4.2µM, respectively).

Formula: C₁₅H₁₀BrN₃O
MW: 328.2 g/mol
MDL Number: MFCD09037522
CAS Number: 676596-65-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3fa502c6ea88f8fc15c893ccc8d097134e80073&vid=87db20f841ae5c1a287473288f4e79216432eebb&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR