Salubrinal ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: (E)-N-(2,2,2-Trichloro-1-(3-(quinolin-8-yl)thioureido)ethyl)cinnamamide

270-428-M001 270-428-M005
89147-876EA 72.68 CAD
89147-876 89147-878
Salubrinal ≥98% (by HPLC)
Salubrinal

Cell permeable and selective inhibitor of the phosphatase complexes that dephosphorylate eukaryotic translation initiation factor 2 subunit alpha (eIF-2alpha). Protects ER stress-induced apoptosis (EC50~15µM in PC12 cells stimulated with 750ng/ml of tunicamycin (Prod. No. BML-CC104)).

Formula: C₂₁H₁₇Cl₃N₄OS
MDL Number: MFCD00548612
CAS Number: 405060-95-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c1b5a39f2162ee796ad89e60aaf2eebec4b3993&vid=4643238a3b4a66766b4db48383254fb81701a96d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR