H-1152 dihydrochloride ≥98% (by HPLC)

Supplier: Enzo Life Sciences

270-423-M001 270-423-M005 270-423-M025
89147-860EA 140.62 CAD
89147-860 89147-862 89147-864
H-1152 dihydrochloride ≥98% (by HPLC)
H-1152 dihydrochloride

A cell permeable, highly specific, potent and ATP-competitive inhibitor of Rho kinase (ROCK) (Ki=1.6nM). Exhibits a much weaker affinity for other serine/threonine kinases (Ki=630nM for PKA, 9.27mM for PKC and 10.1mM for MLCK). More potent and selective than Y-27632. Inhibits the phosphorylation of MARCKS in cells stimulated by lysophosphatidic acid. Inhibits EP3-stimulated NO formation. Prevents fragmentation of apoptotic cells. Relieves neuropathic pain.


  • ROCK inhibitor


White to off-white solid.


Soluble in water.

Formula: C₁₆H₂₁N₃O₂S·2HCl
Storage Temperature: Freezer
CAS Number: 45146-2-58-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e38f398f6350a9037d8e44fb296ce48e346a4bdb&vid=c60e36a36a367bcf21f08ea19b61016228f80496&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR