CVT-313 ≥98% (by HPLC)

Supplier: Enzo Life Sciences

270-393-M001 270-393-M005
89147-790EA 150.1 CAD
89147-790 89147-792
CVT-313 ≥98% (by HPLC)
CVT-313

CDK/cyclin inhibitor


Specific inhibitor of CDK2/cyclin A (IC50=500nM) and CDK2/cyclin E (IC50=500nM). Also inhibits CDK1/cyclin B (IC50=4.2µM) and CDK4/cyclin D1 (IC50=215µM).

Formula: C₂₀H₂₈N₆O₃
MDL Number: MFCD06411412
CAS Number: 199986-75-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4fa4d97c6579a0a60ff90923eda3ab797b527b55&vid=347028670c368893b3cb27e083ecb6c79f620c08&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR