Pioglitazone ≥97% (by HPLC)

Supplier: Enzo Life Sciences
Warning

270-367-M001 270-367-M005
89147-718EA 105.86 CAD
89147-718 89147-720
Pioglitazone ≥97% (by HPLC)
Pioglitazone

PPARgamma activator.


  • Formulation: White to off-white solid.


Pioglitazone selectively activates PPARγ-1. It is about one tenth as potent as rosiglitazone (EC₅₀~500 nM for human and mouse PPARγ). In a transactivation assay using COS-1 cells transfected with full length human PPARα and RXRα, pioglitazone and rosiglitazone exhibit low level activation of PPARα at 1 µM and 5.4- and 4.2-fold activation, respectively, at a concentration of 10 µM.


Soluble in DMSO (2.5 mg/mL) or dimethyl formamide (2.5 mg/mL).

Formula: C₁₉H₂₀N₂O₃S
Storage Temperature: Freezer
MDL Number: MFCD00865504
CAS Number: 111025-46-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e80df1b95ec52c2de58d5a3453031297a5036a9f&vid=a85366a405272892459dbbc271f44589af118343&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥97% (HPLC)
Identity Confirmed by ¹H-NMR

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR