BEC ammonium salt ≥95% (by TLC)

Supplier: Enzo Life Sciences

270-345-M001 270-345-M005
89147-660EA 104.28 CAD
89147-660 89147-662
BEC ammonium salt ≥95% (by TLC)
BEC ammonium salt

Slow-binding competitive inhibitor of arginase I (Ki=0.4-0.6µM & KD=2.22µM for recombinant rat liver arginase) and arginase II (Ki=0.31µM at pH 7.5 and 0.03µM at pH 9.5 for human recombinant type II arginase). Enhances NO-dependent smooth muscle relaxation in human penile corpus cavernosum tissue. Does not inhibit nitric oxide synthase (NOS) and may serve as a valuable reagent to probe the physiological relationship between arginase and NOS. Inhibitor of rat aortic smooth muscle cell proliferation.


  • Arginase inhibitor


White to off-white powder.


Soluble in water (10mg/ml).

Formula: C₅H₁₂BNO₄S·NH₄
Storage Temperature: Refrigerator
CAS Number: 63107-40-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14fec8cbc15d7c8eb872d5d164af28269dd398c9&vid=52628f64d4e5638c655bb4623328cec8564495be&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR