T0901317 ≥98% (by TLC)

Supplier: Enzo Life Sciences

270-309-M010 270-309-M050
89147-616EA 74.26 CAD
89147-616 89147-618
T0901317 ≥98% (by TLC)
T0901317

LXR activator


Potent agonist (EC50=50nM) of the orphan nuclear hormone receptor (liver X receptor; LXR). Inhibitor of human sex hormone-dependent cancer cell lines.

Formula: C₁₇H₁₂F₉NO₃S
MDL Number: MFCD03412028
CAS Number: 293754-55-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b95d727989239acec47c4b261f3040dea527bf70&vid=e1f6d46129a83b28f59a6ed4cc167508753ade5c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR