Sirtinol ≥97% (by NMR)

Supplier: Enzo Life Sciences

Synonyms: 2-(((2-Hydroxynaphthalen-1-yl)methylene)amino)-N-(1-phenylethyl)benzamide

270-308-M001 270-308-M005
89147-612EA 71.1 CAD
89147-612 89147-614
Sirtinol ≥97% (by NMR)
Sirtinol

SIRT inhibitor


Specific cell permeable inhibitor of the sirtuin family of NAD-dependent deacetylases (ySir2: IC50=48µM; hSIRT1: IC50=60µM; hSIRT2: IC50=58µM) with no effect on human HDAC1. Reported to inhibit Sir2p transcriptional silencing activity in vivo (IC50=25µM) and NAD-dependent histone deacetylase activity of purified recombinant yeast Sir2p (IC50=70µM) and hSIRT2 (IC50=40µM) in vitro.

Formula: C₂₆H₂₂N₂O₂
MDL Number: MFCD00810186
CAS Number: 410536-97-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ef03cc03e424f774dda7814bd97ead40ebdedd8&vid=6cc185b4a0feae94919c48a5a24274d128123086&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR