4-(Dimethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide ≥98% (by HPLC)

Supplier: Enzo Life Sciences

270-297-M001 270-297-M005
89147-596EA 237 CAD
89147-596 89147-598
4-(Dimethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide ≥98% (by HPLC)
4-(Dimethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide

Potent inhibitor of histone deacetylases (HDACs) (IC50≤1µM).


  • HDAC inhibitor


White crystalline solid.


Soluble in DMSO (1mg/ml)

Formula: C₁₆H₂₅N₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD03453554
CAS Number: 251456-60-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d893d16e7f9eeb91400f4e93fd152783e75a1905&vid=82ca3ee3071011267c367e3958064b0ebf6bbdb8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR