PGP-4008 ≥93%

Supplier: Enzo Life Sciences

270-290-M002 270-290-M010
89147-580EA 284.4 CAD
89147-580 89147-582
PGP-4008 ≥93%
PGP-4008

Selective P-glycoprotein (Pgp) inhibitor. Effectively reverses Pgp mediated drug resistance in NCI/ADR cells, but has no effect on the drug sensitivities of MCF-7/VP cells that overexpress MRP1.


  • P-glycoprotein inhibitor


White to off-white powder.


Soluble in methanol, 100% ethanol (3mg/ml) or DMSO (20mg/ml).

Formula: C₂₆H₂₃N₃O
Storage Temperature: Freezer
CAS Number: 365565-02-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5307b516330865542e64c64cdb5327d12d7af4c3&vid=9c361a00ca23f70dc0da41643217ffd1f4828126&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR