C75 ≥98%

Supplier: Enzo Life Sciences

270-286-M001 270-286-M005
89147-564EA 153.26 CAD
89147-564 89147-566
C75 ≥98%
C75

Fatty acid synthase inhibitor


Inhibitor of fatty acid synthase (FAS) reducing food intake and body weight in mice. Exhibits irreversible slow-binding biphasic inactivation of fatty acid synthase. Downregulates neuropeptide Y and Agouti-related protein expression. Has been proposed to activate CPT-1 activity in liver and adipose tissue, leading to increased fatty acid oxidation and energy production. Shows significant in vivo antitumor activity in human breast cancer cells. Suppresses DNA replication and induces apoptosis. FAS inhibition by C75 leads to a dramatic accumulation of the CDK inhibitor p27KIP1 from cytosol to cell nuclei.


Synthetic.

Formula: C₁₄H₂₂O₄
MDL Number: MFCD03265452
CAS Number: 191282-48-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1728f9adb02243fa85434bd9aaeaa075c94dd3e&vid=ffa8ec9053282ad942f91f5da47f30769eed2e5b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR