DL-α-Difluoromethylornithine hydrochloride monohydrate ≥98% (by TLC)
Supplier: Enzo Life Sciences
270-283-M010 270-283-M050
89147-556EA 75.84 CAD
89147-556 89147-558
DL-α-Difluoromethylornithine hydrochloride monohydrate ≥98% (by TLC)
DL-α-Difluoromethylornithine hydrochloride monohydrate
Inhibitor of ornithine decarboxylase and arginase
Specific, irreversible inhibitor of ornithine decarboxylase (ODC), the rate-limiting enzyme in polyamine biosynthesis. Potent chemopreventive agent. Induces apoptosis. Arginase activity inhibitor.
CAS Number: 68278-23-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0dc40d4c0bc1409d60278edcd94293b840eeefe&vid=a1138d901434e70b624938ea8d239212f4f72e4f&prid=web&clid=SAPDAM