Daphnetin ≥97%

Supplier: Enzo Life Sciences

Synonyms: 7,8-Dihydroxy-2H-chromen-2-one, 7,8-Dihydroxycoumarin

270-281-M005 270-281-M025
89147-552EA 69.52 CAD
89147-552 89147-554
Daphnetin ≥97%
Daphnetin

Protein kinase inhibitor


Inhibitor of protein kinases. EGF receptor tyrosine kinase (IC50=7.87µM), PKA (IC50=9.33µM), PKC (IC50=25.01µM). Antioxidant. Antimalarial agent.

Formula: C₉H₆O₄
MW: 178.14 g/mol
MDL Number: MFCD00016977
CAS Number: 486-35-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=195dbcc4c99ac0fe88774bf7e0985a33f972b19c&vid=e5f434a867f2017af041f16ee506613d1b5088a4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR