SB202190 hydrochloride ≥97%

Supplier: Enzo Life Sciences

Synonyms: SB-202190 hydrochloride

270-268-M001 270-268-M005
89147-530EA 175.38 CAD
89147-530 89147-532
SB202190 hydrochloride ≥97%
SB202190 hydrochloride

p38 inhibitor


A water soluble form of the potent p38 (SAPK2a) inhibitor SB202190 (Prod. No. BML-EI294).

Formula: C₂₀H₁₄FN₃O·HCl
MW: 367.9 g/mol
CAS Number: 350228-36-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b727442261e3b21cfab018f81ec1130172ed4145&vid=426958d8e703b45641200ec33adc08e4b6956a36&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR