Rosmarinic acid ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: (R,E)-3-(3,4-Dihydroxyphenyl)-2-((3-(3,4-dihydroxyphenyl)acryloyl)oxy)propanoic acid

270-253-M010 270-253-M050
89147-494EA 69.52 CAD
89147-494 89147-496
Rosmarinic acid ≥98% (by HPLC)
Rosmarinic acid

Naturally occurring polyphenolic compound with antioxidant and anti-inflammatory properties.Inhibits several complement-dependent inflammatory processes via inhibition of the C5 convertase. Anticarcinogenic. Inhibitor of lipid peroxidation, TCR-induced T cell activation and proliferation.


Isolated from Rosmarinus officinalis.

Formula: C₁₈H₁₆O₈
MW: 360.318 g/mol
MDL Number: MFCD00017740
CAS Number: 20283-92-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=156f7314e1637f50fd1cc18f7f491bf50fd8ec54&vid=302ecc0ed39b0748ed07f265f9878dcbc38e3378&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR