Aminopurvalanol A ≥97% (by 1H-NMR)

Supplier: Enzo Life Sciences

270-249-M001 270-249-M005
89147-486EA 109.02 CAD
89147-486 89147-488
Aminopurvalanol A ≥97% (by 1H-NMR)
Aminopurvalanol A

CDK/cyclin inhibitor


Cell permeable, reversible and competitive inhibitor of CDK1/cyclin B (IC50=33nM), CDK2/cyclin A (IC50=33nM), CDK2/cyclin E (IC50=28nM) and CDK5/p35 (IC50=20nM).

Formula: C₁₉H₂₆ClN₇O
MDL Number: MFCD06798384
CAS Number: 220792-57-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fb2c986e9bb241de051725a1fae6f5732739ffb&vid=17d093f9823c0a64a948149b3bc17489e325a69a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR