Purvalanol A ≥99% (by NMR)

Supplier: Enzo Life Sciences

Synonyms: 6-[(3-Chloro)anilino]-2(1R)-(isopropyl-2-hydroxyethylamino)-9-isopropylpurine, (R)-2-((6-((3-Chlorophenyl)amino)-9-isopropyl-9H-purin-2-yl)amino)-3-methylbutan-1-ol

270-246-M001 270-246-M005
89147-480EA 124.82 CAD
89147-480 89147-482
Purvalanol A ≥99% (by NMR)
Purvalanol A

Inhibitor of human CDK1 (IC50=4nM), CDK2/cyclin A (IC50=70nM), Cdc2/cyclin B, CDK2/cyclin E (IC50=35nM), CDK4/cyclin D1 (IC50=850nM) as well as CDK5/p35 (IC50=75nM). More membrane permeable than purvalanol B.


  • CDK/cyclin inhibitor


Off-white to yellow powder.


Soluble in DMSO, methanol or dichloromethane.

Formula: C₁₉H₂₅N₆OCl
Storage Temperature: Freezer
MDL Number: MFCD02179211
CAS Number: 212844-53-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ac2f8d3cf7e708e53773a88414e397c048f218f&vid=f5de4690ace6831bac0af6f9fa5f76762f1a7ead&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR