DPQ ≥98% (by HPLC)

Supplier: Enzo Life Sciences

270-221-M001 270-221-M005
89147-418EA 124.82 CAD
89147-418 89147-420
DPQ ≥98% (by HPLC)
DPQ

PARP-1 inhibitor


Very potent poly(ADP-ribose) polymerase-1 (PARP-1) inhibitor.

Formula: C₁₈H₂₆N₂O₂
MDL Number: MFCD01866274
CAS Number: 129075-73-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e555fa032cedc95ff204e4211bae15149a68945&vid=e32a2e184ac6e0d3ca9226b93fb88011012ed6db&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR