Captopril ≥98%
Supplier: Enzo Life Sciences
Synonyms:
(S)-(-)-1-(3-Mercapto-2-methyl-1-oxopropyl)-L-proline, (S)-(-)-1-(3-Mercapto-2-methylpropionyl)-L-proline, (S)-1-((S)-3-mercapto-2-methylpropanoyl)pyrrolidine-2-carboxylic acid
270-212-G001
270-212-G005
89147-398EA
96.38
CAD
89147-398
89147-400
Captopril ≥98%
Captopril
Inhibitor of ACE and leukotriene hydrolase
First orally active angiotensin-converting enzyme (ACE) inhibitor. IC50=2.14nM. Reversible and competitive inhibitor of LTA4 hydrolase (IC50=11µM). Inhibits angiogenesis. Antihypotensive agent.
Formula:
C₉H₁₅NO₃S MW: 217.28 g/mol |
MDL Number:
MFCD00168073 CAS Number: 62571-86-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...