NS-2028 ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: 4H-8-Bromo-1,2,4-oxadiazolo(3,4-d)benz(b)(1,4)oxazin-1-one

270-210-M001 270-210-M005
89147-388EA 82.16 CAD
89147-388 89147-390
NS-2028 ≥98% (by TLC)
NS-2028

A potent and selective, cell-permeable inhibitor of nitric oxide (NO)-sensitive soluble guanylyl cyclase (sGC). Inhibition is irreversible. Inhibits also the basal, the YC-1 and the coenhanced sGC activity. Complete inhibition of NO-induced relaxation in vascular and nonvascular smooth muscle at 1μM.


  • Guanylyl cyclase inhibitor


White to off-white crystalline powder.


Soluble in DMSO or 100% ethanol; insoluble in water.

Formula: C₉H₅BrN₂O₃
Storage Temperature: Freezer
CAS Number: 204326-43-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d788e156e5d398598e42075c991320a7f9f562f2&vid=881de6a7a99f9203bfe125c4b583e34a049bfd50&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR