Piceatannol ≥98%
Supplier: Enzo Life Sciences
Synonyms:
(E)-4-(3,5-Dihydroxystyryl)benzene-1,2-diol
270-202-M001
270-202-M010
270-202-M050
89147-358EA
79
CAD
89147-358
89147-360
89147-362
89147-364
Piceatannol ≥98%
Piceatannol
Syk inhibitor
Originally isolated from Euphorbia lagascae. Selective protein tyrosine kinase Syk inhibitor (IC50~10 µM). In RBL-2H3 cells the selective inhibition of Syk by piceatannol results in inhibition of FcεR1-mediated signaling. Activator of human deacetylase SIRT1.
Synthetic.
SIRT1 activator.
Formula:
C₁₄H₁₂O₄ MW: 244.2427 g/mol |
MDL Number:
MFCD00221715 CAS Number: 10083-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...