Toluene-α-sulfonyl fluoride ≥99% (by GC-FID)

Supplier: Enzo Life Sciences

Synonyms: PMSF, Phenylmethanesulphonyl fluoride, Phenylmethylsulphonyl fluoride, α-Toluenesulfonyl fluoride, α-Toluenesulphonyl fluoride, Phenylmethanesulfonyl fluoride, Phenylmethylsulfonyl fluoride

270-184-G005 270-184-G025
89147-336EA 121.66 CAD
89147-336 89147-338
Toluene-α-sulfonyl fluoride ≥99% (by GC-FID)
Toluene-α-sulphonyl fluoride

Irreversible inhibitor of serine proteases. Widely used fatty acid amid hydrolase (FAAH) inhibitor for pretreatment of brain membrane preparations when testing CB1 receptor activity. Spleen membranes used for CB2 studies do not require this pretreatment, as FAAH activity is absent in these cells. Inhibits also cysteine proteases like papain (reversible by DTT treatment, Prod. No. ALX-280-001), as well as internucleosomal DNA fragmentation in immature thymocytes. For a related, more stable inhibitor see AEBSF.


  • Inhibitor of serine proteases and FAAH


White to off-white needle crystals.


Soluble in 100% ethanol, methanol or isopropanol; insoluble in water.

Formula: C₇H₇FO₂S
MW: 174.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007424
CAS Number: 329-98-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78cae7f83bb7637d14416d57ef4e47dd81881d65&vid=ab31f6835cc729b11650156a43aab9f3cf4baf2a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR