Patulin ≥98% (by HPLC)

Supplier: Enzo Life Sciences

270-111-M001 270-111-M005 270-111-M025
89147-224EA 77.42 CAD
89147-224 89147-226 89147-228
Patulin ≥98% (by HPLC)
Patulin

Inhibitor of protein farnesylation in a cell free assay. Inhibits incorporation of tritiated mevalonate into proteins in whole cells. Mycotoxin with anti-bacterial, potassium uptake inhibitory and possibly carcinogenic activities. Known contaminant of spoiled apples.


Isolated from Penicillium expansum.

Formula: C₇H₆O₄
MDL Number: MFCD00005858
CAS Number: 149-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb7746101fcf545702537c3b96fc16d997cbfd83&vid=d1a4084e228ea4130c2d8d9f146226b00776dc42&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR