1400W dihydrochloride ≥98%

Supplier: Enzo Life Sciences

270-073-M005 270-073-M025
89147-170EA 121.66 CAD
89147-170 89147-172
1400W dihydrochloride ≥98%
1400W dihydrochloride

iNOS inhibitor


A slow, tight binding and highly selective inhibitor of inducible nitric oxide synthase (iNOS/NOS II) in vitro and in vivo. Binds tightly to iNOS with a Kd of 7nM. In contrast, inhibition of nNOS and eNOS is weaker and rapidly reversible with Ki values of 2 and 50µM respectively. Inhibits tumor growth in vivo. Prevents circulatory failure but not organ injury induced by endotoxin. Displays protective effects in oxygen-deprived brain slices.

Formula: C₁₀H₁₅N₃·2HCl
MDL Number: MFCD03428622
CAS Number: 214358-33-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bbc300c8e32ec08731309f1def758c9ae7d21ca&vid=6adddf31a609f43c81ddabc984485b1e8663e76b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR