Cantharidin ≥98% (by TLC)

Supplier: Enzo Life Sciences

270-063-M025 270-063-M100
89147-162EA 127.98 CAD
89147-162 89147-164
Cantharidin ≥98% (by TLC)
Cantharidin

PP2A inhibitor


Potent protein phosphatase 2A (PP2A) inhibitor. At higher concentrations, also inhibits protein phosphatase 1 (PP1).

Formula: C₁₀H₁₂O₄
MDL Number: MFCD00134968
CAS Number: 56-25-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85e700e664f024df7325dd99b9b3127e32829e49&vid=27a4d92605c408dfe352f5307e4681998b21b433&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR