Bisindolylmaleimide III ≥98%

Supplier: Enzo Life Sciences

Synonyms: 3-(1-(3-Aminopropyl)-1H-indol-3-yl)-4-(1H-indol-3-yl)-1H-pyrrole-2,5-dione

270-051-M001 270-051-M005
89147-128EA 96.38 CAD
89147-128 89147-130
Bisindolylmaleimide III ≥98%
Bisindolylmaleimide III

PKC inhibitor


Potent and selective inhibitor of protein kinase C (PKC).

Formula: C₂₃H₂₀N₄O₂
MW: 384.4 g/mol
MDL Number: MFCD01076633
CAS Number: 137592-43-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1081f00450bdf629837947a142c2db63cd5fa2b&vid=03cb293e1e26ef91fb955f9349ea51d46529e7a7&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR