Bisindolylmaleimide II ≥97% (by 1H-NMR)
Supplier: Enzo Life Sciences
270-050-M001
270-050-M005
89147-124EA
105.86
CAD
89147-124
89147-126
Bisindolylmaleimide II ≥97% (by 1H-NMR)
Bisindolylmaleimide II
PKC inhibitor
Formula:
C₂₇H₂₆N₄O₂ MW: 438.5 g/mol |
CAS Number:
137592-45-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...