3-Aminobenzamide ≥97%

Supplier: Enzo Life Sciences

270-044-G001 270-044-G005
89147-114EA 118.5 CAD
89147-114 89147-116
3-Aminobenzamide ≥97%
3-Aminobenzamide

PARP inhibitor


Inhibits endogenous poly-ADP-ribosyltransferases often present in membrane preparations with minimal effect of bacterial toxin mediated ADP-ribosylation. Inhibits nitric oxide-induced apoptosis but not differentiation in HL-60 cells. Protects cells from oxygen radical and nitric oxide toxicity. Inhibits stress-induced apoptosis.

Formula: C₇H₈N₂O
MDL Number: MFCD00007989
CAS Number: 3544-24-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=741851e0f4f1c9c4e4478da291189e4ca363c9d3&vid=e04bb71e9b9844652f9634365d32efaf91bbab3f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR