L-NIL dihydrochloride ≥97%
Supplier: Enzo Life Sciences
270-010-M010 270-010-M050
89147-036EA 93.22 CAD
89147-036 89147-038
L-NIL dihydrochloride ≥97%
L-NIL dihydrochloride
NOS inhibitor
Selective inhibitor of inducible nitric oxide synthase (iNOS; NOS II). Shows anti-inflammatory activity.
CAS Number: 159190-45-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c93eb9dd38e2ccf95110bc8c9132b1d3441f974f&vid=2b74a660cea9c1cd869034169a62649c0a2eb52f&prid=web&clid=SAPDAM