Z-VRPR-FMK
Supplier: Enzo Life Sciences
260-166-M001
260-166-MC05
89147-010EA
1140.76
CAD
89147-010
89147-012
Z-VRPR-FMK
Proteins and Peptides
MALT1 inhibitor
The proteolytic activity of the paracaspase MALT1 is key to NF-κB and T cell activation. Z-VRPR-FMK is a selective MALT1 inhibitor of such proteolytic activity.