Z-IETD-Fmk
Supplier: Enzo Life Sciences
260-144-R020 260-144-R100
89146-960EA 399.74 CAD
89146-960 89146-962
Z-IETD-Fmk
Proteins and Peptides
Caspase and granzyme inhibitor
Irreversible, cell permeable inhibitor of caspase-8 and granzyme B. Inhibits also caspase-10. The inhibitor is designed as a methyl ester to facilitate cell permeability. 