Ac-VEID-AFC
Supplier: Enzo Life Sciences
260-111-M005 260-111-M010
89146-860EA 380.78 CAD
89146-860 89146-862
Ac-VEID-AFC
Proteins and Peptides
Caspase-6 inhibitor
Fluorogenic substrate for caspase-6 (Km=30μM; Kcat/Km=168000 M-1sec-1) and related cysteine proteases. Sequence is based on lamin A site of cleavage.