AC-YVAD-AFC
Supplier: Enzo Life Sciences
260-108-M005
260-108-M010
89146-848EA
249.64
CAD
89146-848
89146-850
AC-YVAD-AFC
Proteins and Peptides
Substrate for caspase-1 and -4
Fluorogenic substrate for caspase-1 and -4.