Protease Inhibitor I aldehyde
Supplier: Enzo Life Sciences
260-089-M001
260-089-M005
89146-810EA
295.46
CAD
89146-810
89146-812
Protease Inhibitor I aldehyde
Proteins and Peptides
Cell permeable inhibitor of the 26S proteasome (MCP; multicatalytic proteinase complex) in HT4 cells. Causes accumulation of ubiquitinylated proteins in neuronal cells. Prevents the activation of NF-κB in response to TNF-α or okadaic acid through inhibition of IκBα degradation.
White powder.
Soluble in 100% ethanol (30mg/ml), DMSO (30mg/ml) or methanol (50mg/ml).