AC-LEHD-PNA
Supplier: Enzo Life Sciences
260-081-M001 260-081-M005
89146-780EA 121.66 CAD
89146-780 89146-782
AC-LEHD-PNA
Proteins and Peptides
Caspase-9 substrate
Chromogenic substrate for caspase-9 and cysteine proteases. Similar to Ac-LEHD-AMC (Prod. No. ALX-260-080) and Ac-LEHD-AFC (Prod. No. ALX-260-116) but cleavage is monitored colorimetrically by absorbance at 405nm.