AC-LEHD-AMC
Supplier: Enzo Life Sciences
260-080-M001 260-080-M005
89146-776EA 104.28 CAD
89146-776 89146-778
AC-LEHD-AMC
Proteins and Peptides
Caspase-9 substrate
Fluorogenic substrate for caspase-9 and related cysteine proteases. The LEHD sequence is derived from combinatorial experiments. Ex.:340-360nm, Em.:440-460nm.
Order Now
SPECIFICATIONS
Protein/peptide name AC-LEHD-AMC AC-LEHD-AMC
Environmentally Preferable
Species
Source
Purity >98%
Peptide content 75-95%