Z-D(OME)E(OME)VD(OME)-Fmk
Supplier: Enzo Life Sciences
260-072-M001
260-072-M005
89146-748EA
628.84
CAD
89146-748
89146-750
Z-D(OME)E(OME)VD(OME)-Fmk
Proteins and Peptides
Inhibitor of caspase-3 and -7
Irreversible, cell permeable inhibitor of caspase-3 and -7.