N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone ≥95% (by TLC)

Supplier: Enzo Life Sciences

260-071-M001 260-071-M005
89146-744EA 560.9 CAD
89146-744 89146-746
N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone ≥95% (by TLC)
N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone

Irreversible, cell permeable broad-spectrum caspase inhibitor.


  • Caspase inhibitor


White to off-white powder.


Soluble in DMSO or dimethyl formamide.

Formula: C₁₁H₁₈FNO₅
Storage Temperature: Freezer
MDL Number: MFCD03453073
CAS Number: 187389-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR