AC-VEID-AMC
Supplier: Enzo Life Sciences
260-064-M001 260-064-M005
89146-736EA 167.48 CAD
89146-736 89146-738
AC-VEID-AMC
Proteins and Peptides
Caspase-6 substrate
Fluorogenic substrate for caspase-6 (Km=30µML, Kcat/Km=168000M-1sec-1) and related cysteine proteases. Sequence is based on lamin A site of cleavage. Ex.:340-360nM, Em.:440-460nm.