AC-WEHD-AMC
Supplier: Enzo Life Sciences
260-057-M001
260-057-M005
89146-712EA
189.6
CAD
89146-712
89146-714
AC-WEHD-AMC
Proteins and Peptides
Substrate for caspase-1 and -5
Fluorogenic substrate for caspase-1 (Km=4µM) and -5 (Km=15µM). Also a substrate for caspase-4 (Km=31µM). This fluorogenic caspase-1 substrate with an optimal tetrapeptide sequence is cleaved 50-fold more efficiently than Ac-YVAD-AMC (Prod. No. ALX-260-024).