AC-IETD-PNA
Supplier: Enzo Life Sciences
260-045-M001
260-045-M005
89146-678EA
202.24
CAD
89146-678
89146-680
AC-IETD-PNA
Proteins and Peptides
Caspase and granzyme substrate
Chromogenic substrate for caspase-6, -8 and granzyme B. Also a substrate for caspase-10. Similar to Ac-IETD-AMC and Ac-IETD-AFC but cleavage is monitored colorimetrically by absorbance at 405 nm.