Ac-IETD-CHO
Supplier: Enzo Life Sciences
260-043-M001 260-043-M005
89146-670EA 156.42 CAD
89146-670 89146-672
Ac-IETD-CHO
Proteins and Peptides
Caspase and granzyme inhibitor
Reversible inhibitor of caspase-6, -8 and granzyme B. Also inhibits caspase-10.