Furin Convertase Substrate (fluorogenic)
Supplier: Enzo Life Sciences
260-040-M001
260-040-M005
89146-662EA
102.7
CAD
89146-662
89146-664
Furin Convertase Substrate (fluorogenic)
Proteins and Peptides
Fluorogenic substrate for furin/SPC1/PACE/PCSK3 and other subtilisin/kexin-like proprotein convertases such as Kex2, SPC2/PC2/PCSK2, SPC3/PCSK1, PACE4/SPC4/PCSK6, SPC5/PC4/PCSK4, SPC6/PC5/PC6/PCSK5, and SPC7/LPC/PC7/PC8/PCSK7. Can be used on live cells. Ex: 340-360 nm, Em: 440-460 nm.