Biotinyl-DEVD-CHO
Supplier: Enzo Life Sciences
260-034-M001 260-034-M005
89146-650EA 225.94 CAD
89146-650 89146-652
Biotinyl-DEVD-CHO
Proteins and Peptides
Inhibitor of caspase-3 and -7
Reversible inhibitor of caspase-3 and -7. Useful for the purification or extract depletion of caspase-3 in conjunction with streptavidin conjugates.