Ac-YVAD-CHO
Supplier: Enzo Life Sciences
260-027-M001
260-027-M005
89146-622EA
132.72
CAD
89146-622
89146-624
Ac-YVAD-CHO
Proteins and Peptides
Inhibitor of caspase-1 and -4
Potent and reversible inhibitor of caspase-1. Also inhibits caspase-4. Strongly inhibits anti-Fas induced apoptosis in L929-Fas cells.