AC-YVAD-PNA
Supplier: Enzo Life Sciences
260-026-M001 260-026-M005
89146-618EA 91.64 CAD
89146-618 89146-620
AC-YVAD-PNA
Proteins and Peptides
Caspase-1 substrate
Sequence is a modification of pro-interleukin-1beta Asp-116 cleavage site for caspase-1. Similar to Ac-YVAD-AMC but cleavage is monitored colorimetrically at 405nm.