Ac-YVAD-AMC
Supplier: Enzo Life Sciences
260-024-M001 260-024-M005
89146-614EA 99.54 CAD
89146-614 89146-616
Ac-YVAD-AMC
Proteins and Peptides
Caspase-1 inhibitor
Fluorgogenic caspase-1 substrate with a Km=14µM. Ex.:340-360nm, Em.:440-460nm.Sequence is based on precursor interleukin-1beta Asp116 cleavage site for caspase-1.