Ac-YVAD-2,6-dimethylbenzoyloxymethylketone
Supplier: Enzo Life Sciences
260-016-M001 260-016-M005
89146-590EA 194.34 CAD
89146-590 89146-592
Ac-YVAD-2,6-dimethylbenzoyloxymethylketone
Proteins and Peptides
Inhibitor of caspase-1 and -4
Irreversible, cell permeable inhibitor of caspase-1. Also inhibits caspase-4.