Amastatin Hydrochloride
Supplier: Enzo Life Sciences
260-003-M001
260-003-M005
89146-558EA
115.34
CAD
89146-558
89146-560
Amastatin Hydrochloride
Proteins and Peptides
Aminopeptidase inhibitor.
Competitive inhibitor of human serum aminopeptidase A, pig kidney leucine aminopeptidase, leucine aminopeptidase, tyrosine aminopeptidase, but not of aminopeptidase B. Has been used to potentiate the action of some bioactive peptides by inhibiting their degradation by aminopeptidases.